Learn More
Medicom™ AssureMask Precision™ Tie-On Face Masks
Catalog No. p-7237328 Shop All Medicom Products
This item is not returnable. View return policy
Catalog No. 17-000-563 Supplier Medicom™ Supplier No. 206815
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Tie-on face masks provide ASTM Level 1, 2 or 3 protection. Made from high quality materials that enhance breathability, these masks have soft, ample-length ties, an adjustable nose piece and a soft inner lining to ensure a comfortable, customized fit.
  • Non-woven spunbond outer layer, provides maximum protection against fluid penetration
  • Ample length ties are soft, strong, ultrasonically sealed and attached to the outside of the mask to prevent pulling and irritation
  • Shingle pleat design prevents fluid pooling and cross-contamination
  • High quality media ensures optimal filtration and breathability
  • SureView™ Technology Fog-free nose strip ensures an unobstructed view of the operative field
Specifications
Product Type Medical/Procedural Face Mask
Fastening Type Ties
Material Spunbond Polypropylene
Size Standard
Sterility Non-sterile
Style Pleated
Disposable Single-use
Filter Efficiency ≥95% (PFE), ≥95% (BFE)
Latex Free Yes
Model 206815
Packaging Quantity 50 Masks/Box, 6 Boxes/Cs.
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov