Learn More
Medicom™ AssureMask Valiant™ Earloop Face Masks
Procedure earloop face masks provide ASTM Level 1, 2 or 3 protection as well as optimal comfort and breathability.
Supplier:  Medicom™ 205716
 View more versions of this product
Catalog No. 17-000-552
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
  • Non-woven spunbond polypropylene outer layer provides maximum protection against fluid penetration
  • High-quality media ensure optimal filtration and breathability
  • Inner layer is made of dye-free non-woven spunbond polypropylene that will not lint or tear during use
  • Ample-length earloops are soft, strong, free of natural rubber latex, ultrasonically sealed, and attached to the outside of the mask to prevent pulling and irritation
  • Adjustable, enclosed, plastic-covered, iron nosepiece provides customized protection and comfort
  • Shingle pleat design prevents fluid pooling and cross-contamination
  • Made in North America to meet the highest quality and regulatory standards
  • Available in assorted colors
Specifications
Specifications
Medical/Procedural Face Mask
Spunbond Polypropylene
Non-sterile
Single-use
Yes
50 Masks/Box, 10 Boxes/Cs.
Earloops
Standard
Pleated
≥95% (PFE), ≥95% (BFE)
205716
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.