Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medicom™ AssureTouch™ Agile™ Nitrile Medical Examination Gloves with Textured Fingertips PROMO

Catalog No. 17101085PM
Click to view available options
Size:
Medium
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

These nitrile glove offer optimal puncture and tear resistance without compromising dexterity or tactility.

  • Textured fingertips ensure a secure grip, even when wet
  • Chemo tested

Specifications

Chlorinated No
Accelerator Free No
Applications Healthcare, Industrial, Laboratory
AQL 1.5
Color Blue
Disposable Single-use
Exterior Finish Fingertip Textured
Latex Free Yes
Powdered Powder Free
Size Medium
Sterility Non-sterile
FDA 510 (k) Yes
Glove Cuff Style Beaded
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.