Learn More
MilliporeSigma™ Supelco™ Astec™ CHIROBIOTIC HPLC column Screening Kit, 5 μm particle size
Shop All MilliporeSigma Supelco ProductsDescription
- Material: Stainless steel column
- Pore size: 100 Å
- Matrix: Fully porous particle
- Separation technique: Chiral
- Kit contents: One each of Astec™ CHIROBIOTIC™ V2, T, TAG and R
- Operating temperature: 0 to 45°C
- Operating pressure: 241 bar
- Operating pH: 3.5 to 6.8
- Suitable for: HPLC, LC/MS
- Mobile phase: 1-10 mM copper sulfate (CuSO4)
- Recommended starting mobile phase: 20% methanol in 5 mM copper sulfate
Specifications
Specifications
| Diameter (Metric) | 4.6 mm |
| Length (Metric) | 25 cm |
| Particle Size | 5 μm |
| Pore Size | 100 Å |
| Stationary Phase | Macrocyclic Glycopeptides |
| Product Line | Astec CHIROBIOTIC |
| Phase | Chiral |
| Material | Stainless Steel |
| Technique | Chiral HPLC |
Astec is a registered trademark of Sigma-Aldrich Co. LLC
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.