Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Astec™ CHIROBIOTIC™ R Chiral HPLC Column, 5 μm Particle Size

Catalog No. 111005848 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Length (Metric):
10 cm
15 cm
25 cm
Diameter (Metric):
2.1 mm
4.6 mm
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric)
11-100-5848 10 cm 4.6 mm
11-100-5786 15 cm 2.1 mm
11-100-5755 25 cm 2.1 mm
11-100-5790 15 cm 4.6 mm
11-100-5742 25 cm 4.6 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 11-100-5848 Supplier MilliporeSigma™ Supelco™ Supplier No. 13022AST
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CHIROBIOTIC™ R, based on the ristocetin A glycopeptide covalently bonded to high purity silica particles, has shown particular applicability to enantiomers of anionic compounds.

Selectivity on CHIROBIOTIC™ R strongly correlates to the organic modifier, favoring the alcohol-type mobile phases by a large margin.
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 4.6 mm
Length (Metric) 10 cm
Particle Size 5 μm
pH 3.5 to 6.8
Pore Size 100 Å
Stationary Phase Ristocetin A Glycopeptide
Product Line Astec CHIROBIOTIC
Matrix Ristocetin A Glycopeptide Phase/High-Purity Silica Gel Particle Platform
Phase Chiral
Material Stainless Steel
Technique Chiral HPLC
Show More Show Less

Astec and CHIROBIOTIC are registered trademarks of Sigma-Aldrich Co. LLC.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.