Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Astemizole
SDP

Catalog No. 348950 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-895-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 34-895-0 Supplier Tocris Bioscience™ Supplier No. 3489/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Orally active, potent H1 antagonist. Also KV11.1 (hERG) channel blocker

Orally active, potent histamine H1 antagonist (IC50 = 4 nM) that displays 20-fold, > 250-fold and > 250-fold selectivity over 5-HT, dopamine and muscarinic acetylcholine receptors respectively. Exhibits antimalarial activity in multidrug resistant strains in vitro (IC50 = 227 - 734 nM). Also potent KV11.1 (hERG) channel blocker (IC50 = 0.9 nM) that displays cardiotoxicity in vivo.

Chemical Identifiers

CAS 68844-77-9
Molecular Formula C28H31FN4O

Specifications

Quantity 50 mg
Formula Weight 458.57
Purity >99%
Molecular Weight (g/mol) 458.581
Chemical Name or Material Astemizole
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.