Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ASTL Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144141
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144141 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144141 Supplier Invitrogen™ Supplier No. PA5144141
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ASTL Polyclonal Antibody for IHC (P), ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen ASTL
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene ASTL
Gene Accession No. Q6HA08
Gene Alias astacin-like metalloendopeptidase; astacin-like metalloendopeptidase (M12 family); astacin-like metallo-endopeptidase (M12 family); ASTL; Oocyte astacin; ovastacin; SAS1B; sperm acrosomal SLLP1 binding
Gene Symbols ASTL
Host Species Rabbit
Immunogen Recombinant Human Astacin-like metalloendopeptidase protein (38-337AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 431705
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.