Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ASTN1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP243576

Catalog No. NBP243576


Only null left
Add to Cart

Description

Description

Specifically detects ASTN1 in Mouse samples, and it is validated for Western Blotting

ASTN1 Polyclonal specifically detects ASTN1 in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

ASTN1
Polyclonal
Western Blot 1:500-1:3000
ASTN1
Recombinant protein encompassing a sequence within the center region of human Astrotactin. The exact sequence is proprietary.
Immunogen affinity purified
RUO
460
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Western Blot
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
Rabbit
145 kDa
0.1 mL
Primary
Mouse
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only