Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ASTN2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555491
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555491 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555491 Supplier Invitrogen™ Supplier No. PA555491
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DSKFNDTLFG EMLHGYNNRT QHVNQGQVFQ MTFRENNFIK DFPQLADGLL VIPLPVEEQC RGVLSEPLPD LQLLTGDIRY DEAMGYPMVQ QWRVRSNLYR VKLSTITLAA GFTNVLKILT KESSREELLS FIQHYGSHY Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.

This gene encodes a protein that is expressed in the brain and may function in neuronal migration, based on functional studies of the related astrotactin 1 gene in human and mouse. A deletion at this locus has been associated with schizophrenia. Multiple transcript variants encoding different proteins have been found for this locus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASTN2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ASTN2
Gene Accession No. O75129
Gene Alias 1d8; Astn2; Astnl; astrotactin 2; astrotactin 2-like; astrotactin-2; bA264C15.1; bA67K19.1; bM243L4.1 (astrotactin 2); bM264P11.1 (astrotactin 2); bM383P24.1 (astrotactin 2); bM423P8.1 (astrotactin 2); bM452J22.1; bM452J22.1 (novel protein); bM468K2.1 (astrotactin 2); Kiaa0634
Gene Symbols ASTN2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ASTN2. Recombinant protein control fragment (Product #RP-90988).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23245
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.