Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Astrin Antibody, Biotin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10074638B
Description
Astrin Polyclonal antibody specifically detects Astrin in Human samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
Astrin | |
Polyclonal | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
Astrin, Deepest, hMAP126, MAP126mitotic spindle coiled-coil related protein, mitotic spindle associated protein p126, Mitotic spindle-associated protein p126, sperm associated antigen 5, sperm-associated antigen 5 | |
The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human astrin (mitotic spindle associated protein p126) using the numbering given in entry NP_006452.3 (GeneID 10615). | |
0.1 mL | |
Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, MAP Kinase Signaling, Stem Cell Markers | |
10615 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunofluorescence | |
Biotin | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction