Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Astrocyte Monoclonal Antibody (ASTRO1 (10E4/R5)), Invitrogen™

Catalog No. PIMA191360
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA191360 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA191360 Supplier Thermo Scientific Supplier No. MA191360

Mouse Monoclonal Antibody

MA1-91360 detects Astrocyte from human samples.

Astrocytes are a type of glial cell found in the brain and spinal cord. They hold various roles, primarily supporting neurons. They are the most abundant glial cells in the brain and closely associate with neuronal synapses, regulating the transmission of electrical impulses. Astrocytes function in the biochemical support of endothelial cells that form the blood-brain barrier, provide nutrients to nervous tissue, maintain the extracellular ion balance, and play a role in the repair and scarring process of the brain and spinal cord following traumatic injuries.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Astrocyte
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone ASTRO1 (10E4/R5)
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation 10mM PBS with 0.2% BSA and 0.1% sodium azide; pH 7.4
Gene Alias Astrocytes
Host Species Mouse
Immunogen Proprietary immunogen.
Purification Method Protein A
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage 4° C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.