Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Astrovirus Capsid Monoclonal Antibody (4988)
Astrovirus Capsid Monoclonal Antibody (4988)
Supplier: Thermo Scientific MA547548
Description
Astrovirus Capsid Monoclonal Antibody for ELISA

Specifications
Astrovirus Capsid | |
Monoclonal | |
100 μg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Proprietary sequence. | |
100 μg | |
Primary | |
4°C | |
Liquid |
ELISA | |
4988 | |
Unconjugated | |
Mouse | |
Protein A | |
RUO | |
Virus | |
Antibody | |
IgG2a |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction