Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ASUN Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$353.00 - $573.00
Specifications
Antigen | ASUN |
---|---|
Dilution | Western Blot 0.04 to 0.4 μg/mL, Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL |
Applications | Western Blot, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
ASUN Polyclonal antibody specifically detects ASUN in Human samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
ASUN | |
Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Human | |
ASUN, asunder, spermatogenesis regulator homolog (Drosphila), cell cycle regulator Mat89Bb homolog, chromosome 12 open reading frame 11, FLJ10630, Mat89Bb, NET48, SPATA30 | |
This antibody was developed against a recombinant protein corresponding to the amino acids: RGKEELAEAEIIKDSPDSPEPPNKKPLVEMDETPQVEKSKGPVSLLSLWSNRINTANSRKHQEFAGRLNSVNN | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot 0.04 to 0.4 μg/mL, Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL | |
Polyclonal | |
Purified | |
RUO | |
PBS, pH 7.2, 40% glycerol | |
55726 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title