Learn More
Asunder Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303575A

Description
The recommended shelf life for this product is 1 year from date of receipt.
Crucial regulator of the mitotic cell cycle and development.Specifications
Asunder | |
Polyclonal | |
Unconjugated | |
INTS13 | |
C12orf11, Mat89Bb, NET48, cell cycle regulator Mat89Bb homolog, sarcoma antigen NY-SAR-95 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55726, 71177 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.2 mg/ml | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q8QZV7, Q9NVM9 | |
Ints13 | |
Between 656 and 706 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.