Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ASXL1 Monoclonal Antibody (11I4), Invitrogen™

Catalog No. PIMA536238
Encompass
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA536238 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA536238 Supplier Thermo Scientific Supplier No. MA536238

Mouse Monoclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL.

ASXL1 Monoclonal antibody specifically detects ASXL1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASXL1
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone 11I4
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4MG trehalose and 0.05MG sodium azide
Gene Asxl1
Gene Accession No. P59598, Q8IXJ9
Gene Alias additional sex combs like 1; additional sex combs like 1, transcriptional regulator; additional sex combs like transcriptional regulator 1; additional sex combs-like protein 1; Asxl1; BOPS; KIAA0978; MDS; MGC117280; MGC71111; mKIAA0978; Polycomb group protein ASXL1; putative Polycomb group protein ASXL1; RGD1561878
Host Species Mouse
Immunogen A synthetic peptide corresponding to a sequence of human ASXL1 (KKERTWAEA ARLVLENYSDAPMTPKQILQVIEAE).
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171023, 228790, 311553
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.