Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ASXL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571475
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571475 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571475 Supplier Invitrogen™ Supplier No. PA571475
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ASXL1 Polyclonal Antibody for Flow, ICC/IF, IHC (P), Western Blot

Probable Polycomb group (PcG) protein involved in transcriptional regulation mediated by ligand-bound nuclear hormone receptors, such as retinoic acid receptors (RARs) and peroxisome proliferator-activated receptor gamma (PPARG). Acts as coactivator of RARA and RXRA through association with NCOA1. Acts as corepressor through recruitment of KDM1A and CBX5 to target genes in a cell-type specific manner; the function seems to involve differential recruitment of methylated histone H3 to respective promoters. Acts as corepressor for PPARG and suppresses its adipocyte differentiation-inducing activity. Non-catalytic component of the PR-DUB complex, a complex that specifically mediates deubiquitination of histone H2A monoubiquitinated at 'Lys-119' (H2AK119ub1).
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASXL1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.47 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Asxl1
Gene Accession No. P59598, Q8IXJ9
Gene Alias additional sex combs like 1; additional sex combs like 1, transcriptional regulator; additional sex combs like transcriptional regulator 1; additional sex combs-like protein 1; Asxl1; BOPS; KIAA0978; MDS; MGC117280; MGC71111; mKIAA0978; Polycomb group protein ASXL1; putative Polycomb group protein ASXL1; RGD1561878
Gene Symbols Asxl1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 521-549 amino acids from the Central region of human ASXL1.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171023, 228790
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.