Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ASXL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565126
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565126 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565126 Supplier Invitrogen™ Supplier No. PA565126
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PKSMPVSEASL IRIVPVVSQS ECKEEALQMS SPGRKEECES QGEVQPNFST SSEPLLSSAL NTHELSSILP IKCPKDE Highest antigen sequence identity to the following orthologs - mouse 62%, rat 54%.

Putative Polycomb group (PcG) protein. PcG proteins act by forming multiprotein complexes, which are required to maintain the transcriptionally repressive state of homeotic genes throughout development. PcG proteins are not required to initiate repression, but to maintain it during later stages of development. They probably act via methylation of histones, rendering chromatin heritably changed in its expressibility. Involved in transcriptional regulation mediated by ligand-bound nuclear hormone receptors, such as peroxisome proliferator-activated receptor gamma (PPARG). Acts as coactivator for PPARG and enhances its adipocyte differentiation-inducing activity; the function seems to involve differential recruitment of acetylated and methylated histone H3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASXL2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ASXL2
Gene Accession No. Q76L83
Gene Alias 4930556B16Rik; additional sex combs like 2; additional sex combs like 2 (Drosophila); additional sex combs like 2, transcriptional regulator; additional sex combs like transcriptional regulator 2; additional sex combs-like protein 2; ASXH2; ASXL transcriptional regulator 2; ASXL2; Kiaa1685; mKIAA1685; polycomb group protein ASXH2; polycomb group protein ASXH2 homolog; putative Polycomb group protein ASXL2
Gene Symbols ASXL2
Host Species Rabbit
Immunogen Recombinant Human ASXL2. Recombinant protein control fragment (Product #RP-105887).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55252
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.