Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ASXL2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568361
Description
ASXL2 Polyclonal Antibody for IHC (P)
Putative Polycomb group (PcG) protein. PcG proteins act by forming multiprotein complexes, which are required to maintain the transcriptionally repressive state of homeotic genes throughout development. PcG proteins are not required to initiate repression, but to maintain it during later stages of development. They probably act via methylation of histones, rendering chromatin heritably changed in its expressibility. Involved in transcriptional regulation mediated by ligand-bound nuclear hormone receptors, such as peroxisome proliferator-activated receptor gamma (PPARG). Acts as coactivator for PPARG and enhances its adipocyte differentiation-inducing activity; the function seems to involve differential recruitment of acetylated and methylated histone H3.
Specifications
ASXL2 | |
Polyclonal | |
Unconjugated | |
ASXL2 | |
4930556B16Rik; additional sex combs like 2; additional sex combs like 2 (Drosophila); additional sex combs like 2, transcriptional regulator; additional sex combs like transcriptional regulator 2; additional sex combs-like protein 2; ASXH2; ASXL transcriptional regulator 2; ASXL2; Kiaa1685; mKIAA1685; polycomb group protein ASXH2; polycomb group protein ASXH2 homolog; putative Polycomb group protein ASXL2 | |
Rabbit | |
affinity chromatography | |
RUO | |
55252 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.8 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q76L83 | |
ASXL2 | |
Synthetic protein corresponding to residues near the C terminal of human ASXL2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction