Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ASXL2 Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. ABE1320 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE1320 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE1320 Supplier MilliporeSigma Supplier No. ABE1320
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-ASXL2 is a highly specific rabbit polyclonal antibody that targets Additional sex combs-like protein 2 and has been tested in Immunocytochemistry and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASXL2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols ASXL2,ASXH2,KIAA1685
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to 22 amino acids from the internal region of human Additional sex combs-like protein 2 (ASXL2).
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_060733
Test Specificity This rabbit polyclonal antibody detects Additional sex combs-like protein 2 (ASXL2) in human cells. It targets an epitope within 22 amino acids from the internal region.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.