Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ASY3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144401
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144401 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144401 Supplier Invitrogen™ Supplier No. PA5144401
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ASY3 Polyclonal Antibody for Western Blot, ELISA

Required for normal meiosis in male and female gametophytes. Acts with ASY1 at the interface between the developing chromosome axes and the recombination machinery to ensure interhomolog recombination. Required for synaptonemal complex formation during meiosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASY3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.964 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene ASY3
Gene Accession No. Q0WR66
Gene Alias Meiosis-specific protein ASY3 ASY3 At2g46980
Gene Symbols ASY3
Host Species Rabbit
Immunogen Recombinant Arabidopsis thaliana ASY3 protein (599-793aa).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 819312
Target Species Plant
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.