Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ASZ1 Monoclonal Antibody (3C9)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA524521
Description
Peptide Sequence: QILKCVELLL SRNADPNVAC RRLMTPIMYA ARDGHTQVVA LLVAHGAEVN TQDENGYTAL TWAARQGHKN IVLKLLELGA NKMLQTKDGK MPSEIAKRNK HHEIFNLL.
Plays a central role during spermatogenesis by repressing transposable elements and preventing their mobilization, which is essential for the germline integrity. Acts via the piRNA metabolic process, which mediates the repression of transposable elements during meiosis by forming complexes composed of piRNAs and Piwi proteins and governs the methylation and subsequent repression of transposons. Its association with pi-bodies suggests a participation in the primary piRNAs metabolic process. Required prior to the pachytene stage to facilitate the production of multiple types of piRNAs, including those associated with repeats involved in the regulation of retrotransposons. May act by mediating protein-protein interactions during germ cell maturation.
Specifications
ASZ1 | |
Monoclonal | |
Unconjugated | |
Asz1 | |
4933400N19Rik; ALP1; ANKL1; ankyrin repeat, SAM and basic leucine zipper domain containing 1; ankyrin repeat, SAM and basic leucine zipper domain-containing protein 1; ankyrin-like 1; ankyrin-like protein 1; Asz1; C7orf7; CT1.19; GASZ; germ cell-specific ankyrin, SAM and basic leucine zipper domain containing protein; germ cell-specific ankyrin, SAM and basic leucine zipper domain-containing protein; Orf3 | |
Mouse | |
Protein A | |
RUO | |
136991 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
3C9 | |
PBS with no preservative; pH 7.4 | |
Q8WWH4 | |
Asz1 | |
ASZ1 (NP_570124, 127 a.a. ~ 234 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction