Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ASZ1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988988
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8988 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8988 Supplier Bioss Supplier No. BS9325R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Plays a central role during spermatogenesis by repressing transposable elements and preventing their mobilization, which is essential for the germline integrity. Acts via the piRNA metabolic process, which mediates the repression of transposable elements during meiosis by forming complexes composed of piRNAs and Piwi proteins and governs the methylation and subsequent repression of transposons. Its association with pi-bodies suggests a participation in the primary piRNAs metabolic process. Required prior to the pachytene stage to facilitate the production of multiple types of piRNAs, including those associated with repeats involved in the regulation of retrotransposons. May act by mediating protein-protein interactions during germ cell maturation.

Specifications

Antigen ASZ1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Asz1
Gene Accession No. Q8WWH4
Gene Alias 4933400N19Rik; ALP1; ANKL1; ankyrin repeat, SAM and basic leucine zipper domain containing 1; Ankyrin repeat, SAM and basic leucine zipper domain-containing protein 1; ankyrin-like 1; Ankyrin-like protein 1; ASZ1; C7orf7; CT1.19; Gasz; germ cell-specific ankyrin, SAM and basic leucine zipper domain containing protein; Germ cell-specific ankyrin, SAM and basic leucine zipper domain-containing protein; ORF3
Gene Symbols Asz1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ASZ1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 136991
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.