Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ At5g63020 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144531
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144531 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144531 Supplier Invitrogen™ Supplier No. PA5144531
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

At5g63020 Polyclonal Antibody for ELISA, Western Blot

Probable disease resistance protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen At5g63020
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.49 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene At5g63020
Gene Accession No. Q8RXS5
Gene Alias Probable disease resistance protein At5g63020
Gene Symbols At5g63020
Host Species Rabbit
Immunogen Recombinant Arabidopsis thaliana At5g63020 protein (360-450aa).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 836422
Target Species Plant
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.