Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Worldwide™ Protective Products ATA™ 505 Cut-Resistant Gloves with Nitrile Hybrid Grip Coating

Catalog No. 18701005 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-701-005 Small
18-701-009 2X-Large
18-701-006 Medium
18-701-007 Large
18-701-008 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 18-701-005 Supplier Worldwide™ Protective Products Supplier No. 505S
Add to Cart
Add to Cart
This item is not returnable. View return policy

Lightweight gloves feature ATA™ Fiber Technology for manual dexterity and a natural fit. Nitrile hybrid coating improves grip capabilities and provides ANSI Level 3 Abrasion Resistance.

  • Lightweight, coated cut-resistant gloves feature ATA fiber technology
  • Gloves offer ANSI Level A4 Cut Protection, and ANSI Level 3 Abrasion Resistance
  • Coated with thin, nitrile hybrid palm dip to improve grip and enhance fingertip sensitivity
  • Designed for manual dexterity with a natural fit and easy flexibility

Specifications

ANSI Abrasion Rating ANSI 3
Coating Material Nitrile
Color Natural
Cut Resistance Rating ANSI A4
Hand Specific
Latex Free Yes
Material ATA Engineered Fiber
Product Type Cut-resistant Gloves
Size Small
For Use With (Application) Metal Fabrication, Construction, Glass Handling, Appliance Manufacturing, and Public Utilities
Glove Cuff Style Continuous Knit
Series 505
Yarn Gauge 13
Length (English) 9.5 in.
Length (Metric) 24.1 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.