Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Worldwide™ Protective Products ATA™ ActivGrip 534 Cut-Resistant Gloves with Nitrile MicroFinish™ Coating

Catalog No. 18701004 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-701-004 2X-Large
18-701-000 Small
18-701-001 Medium
18-701-002 Large
18-701-003 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 18-701-004 Supplier Worldwide™ Protective Products Supplier No. 5342XL
This item is not returnable. View return policy

Nitrile MicroFinish™ surface coating features microscopic suction cups that create a vacuum, dispersing oil on contact for enhanced grip capabilities.

  • 13-gauge, ATA coated cut resistant gloves
  • Lightweight design for dexterity, sensitivity, and comfort
  • ANSI A4 Cut Resistance for handling metal, glass, and other sharp edges
  • ActiveGrip Nitrile MicroFinish palm coating enhances grip on dry, wet, and oily surfaces
  • MicroFinish surface features microscopic suction cups that create a vacuum, dispersing oil on contact
  • ANSI Level 4 Abrasion Resistance for durability and extended useful life
  • Sanitized protective finish provides lasting freshness

Specifications

ANSI Abrasion Rating ANSI 4
Coating Material Nitrile
Color Yellow
Cut Resistance Rating ANSI A4
Hand Specific
Material ATA Engineered Fiber
Product Type Cut-resistant Gloves
Size 2X-Large
For Use With (Application) Automotive Assembly, Glass/Metal Handling, Construction, Pulp/Paper, and Appliance Manufacturing
Glove Cuff Style Continuous Knit
Series 534
Yarn Gauge 13
Length (English) 10.2 in.
Length (Metric) 26 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.