Learn More
Worldwide™ Protective Products ATA™ M530 Cut-Resistant, High Dexterity Gloves
Shop All Protective Industrial Products Inc. ProductsDescription
- Uses ATA™ fiber technology to deliver a thin, form-fitting and lightweight design that supplies ANSI A4 cut protection appropriate for medium-to-high cut hazards
- The 13-gauge seamless knit gloves provide the dexterity and fingertip sensitivity you need to pick up broken glass, dissect samples, and work with task-based knives and scalpels
- Wear them under unlined nitrile gloves to add a cut resistant barrier
- The ATA™ fiber blend also uses Lycra fibers for lots of stretch, making gloves comfortable enough for extended periods of wear
- The Hide-Away technology provides long-lasting abrasion resistance
- Color coded hems make it easy to choose the right glove size
Specifications
Specifications
| Coating Material | Uncoated |
| Color | Green |
| Cut Resistance Rating | ANSI A4 |
| Hand | Specific |
| Latex Free | Yes |
| Material | ATA Engineered Fiber |
| Product Type | Cut-resistant Gloves |
| Size | X-Small |
| For Use With (Application) | Glass Handling, Metal Fabrication, Plastics and Molding, Use as a Liner Glove |
| Glove Cuff Color | Orange |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.