Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Worldwide™ Protective Products ATA™ Sta-Cool Plated Hi-Vis Cut-Resistant Gloves

Catalog No. 18701033 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-701-033 Small
18-701-034 Medium
18-701-035 Large
18-701-036 X-Large
18-701-037 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 18-701-033 Supplier Worldwide™ Protective Products Supplier No. MATAULTRA13HVOPD1S
This item is not returnable. View return policy

Lightweight gloves deliver dexterity and fingertip sensitivity while providing ANSI A4 Cut Protection. Sta-Cool Fabric wicks away moisture for dry, comfortable hands.

  • Lightweight gloves feature ATA fiber technology to deliver ANSI A4 Cut Resistance
  • 13-gauge, seamless knit gloves provide dexterity and fingertip sensitivity
  • Plated with Sta-Cool fiber to wick away moisture for dry, comfortable hands
  • Hi-vis orange color increases awareness to reduce the chance of accidents
  • Machine washable for extended use

Specifications

Color Hi-vis Orange
Cut Resistance Rating ANSI A4
Hand Specific
Material ATA Engineered Fiber
Product Type Cut-resistant Gloves
Size Small
For Use With (Application) Glass Handling, Handling Small Assembly, Automotive Repair, Metal Fabrication, and Aerospace
Glove Cuff Style Continuous Knit
Model MATA-ULTRA13HVO-PD1-S
Series MATA-ULTRAHVO13A
Yarn Gauge 13
Length (English) 8.7 in.
Length (Metric) 22.2 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.