Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATAD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558014
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558014 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558014 Supplier Invitrogen™ Supplier No. PA558014
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NVDRHVDLLE VAQETDGFSG SDLKEMCRDA ALLCVREYVN STSEESHDED EIRPVQQQDL HRAIEKMKKS KDAAFQ Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

ATPase that plays a critical role in regulating the surface expression of AMPA receptors (AMPAR), thereby regulating synaptic plasticity and learning and memory. Required for NMDA-stimulated AMPAR internalization and inhibition of GRIA1 and GRIA2 recycling back to the plasma membrane; these activities are ATPase-dependent.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATAD1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ATAD1
Gene Accession No. Q8NBU5
Gene Alias 4921525H23Rik; AAA+ ATPase Thorase; Ab2-088; AFDC1; Atad1; ATPase family AAA domain containing 1; ATPase family AAA domain-containing protein 1; ATPase family, AAA domain containing 1; ATPase Thorase; AW107648; FLJ14600; FNP001; neuroprotective protein 6; Npg6; Thorase
Gene Symbols ATAD1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ATAD1. Recombinant protein control fragment (Product #RP-97093).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84896
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.