Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATAD2 Monoclonal Antibody (CL0182)
GREENER_CHOICE

Catalog No. PIMA534967
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA534967 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA534967 Supplier Invitrogen™ Supplier No. MA534967
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Immunogen sequence: TAYAIIKEEL DEDFEQLCEE IQESRKKRGC SSSKYAPSYY HVMPKQNSTL VGDKRSDPEQ NEKLKTPSTP VACSTPAQLK RKIRKKSNWY LGTIKKRRKI SQAKDDSQNA IDHK Highest antigen sequence indentity to the following orthologs: Mouse - 82%, Rat - 82%.

ATAD2 is a member of the ATPase family of proteins. Members of this family share a conserved region of around 220 amino acids that contain an ATP-binding site. They also contain one or two AAA (ATPases Associated with Diverse Cellular Activities) domains. ATPase family proteins perform chaperone-like functions that assist in assembly, operation, and disassembly of protein complexes. Involved in the estrogen-induced cell proliferation and cell cycle progression of breast cancer cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATAD2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone CL0182
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ATAD2
Gene Accession No. Q6PL18
Gene Alias 2610509G12Rik; AAA nuclear coregulator cancer-associated protein; ANCCA; Atad2; ATPase family AAA domain-containing protein 2; ATPase family, AAA domain containing 2; CT137; L16; PRO2000
Gene Symbols ATAD2
Host Species Mouse
Immunogen Recombinant protein fragment of Human ATAD2 (Product #RP-92425)
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29028
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.