Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATAD2B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583233
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583233 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583233 Supplier Invitrogen™ Supplier No. PA583233
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LEVVSFCDSG DKCSSEQKIL LEDQSKEKPE TSTENHGDDL EKLEALECSN NEKLEPGSDV EVKDAELDKE GASKVKKYRK Highest antigen sequence indentity to the following orthologs: Rat - 63%, Mouse - 69%.

The protein encoded by this gene belongs to the AAA ATPase family. This family member includes an N-terminal bromodomain. It has been found to be localized to the nucleus, partly to replication sites, consistent with a chromatin-related function. Alternative splicing of this gene results in multiple transcript variants. [provided by RefSeq, Jul 2014].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATAD2B
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ATAD2B
Gene Accession No. Q9ULI0
Gene Alias 1110014E10Rik; ANCCA; ATAD2; Atad2b; ATPase family AAA domain containing 2B; ATPase family AAA domain-containing protein 2B; ATPase family, AAA domain containing 2B; BC032887; C79189; D530031C13Rik; KIAA1240; L16; PRO2000
Gene Symbols ATAD2B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ATAD2B. Recombinant protein control fragment (Product #RP-96053).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54454
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.