Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATAD2B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546087
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546087 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546087 Supplier Invitrogen™ Supplier No. PA546087
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: PGAGAEPGAT GGSSHFISSR TRSSKTRAAS CPAAKAGGSG GAGVTLDEAR Sequence homology: Cow: 100%; Dog: 92%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 100%.

The protein encoded by this gene belongs to the AAA ATPase family. This family member includes an N-terminal bromodomain. It has been found to be localized to the nucleus, partly to replication sites, consistent with a chromatin-related function. Alternative splicing of this gene results in multiple transcript variants. [provided by RefSeq, Jul 2014].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATAD2B
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ATAD2B
Gene Accession No. Q9ULI0
Gene Alias 1110014E10Rik; ANCCA; ATAD2; Atad2b; ATPase family AAA domain containing 2B; ATPase family AAA domain-containing protein 2B; ATPase family, AAA domain containing 2B; BC032887; C79189; D530031C13Rik; KIAA1240; L16; PRO2000
Gene Symbols ATAD2B
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human ATAD2B (aa 23-72).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54454
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.