Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATAD3A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596977
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596977 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596977 Supplier Invitrogen™ Supplier No. PA596977
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSWLFGINKG PKGEGAGPPP PLPPAQPGAE GGGDRGLGDR PAPKDKWSNF DPTGLERAAK AARELEHSRY AKDALNLAQM QEQTLQLEQQ SKLKEYEAAV EQLKSEQIRA QAEERRKTLS EETRQHQARA QYQDKLARQR YEDQLKQQQL LNEENLRKQE ESVQKQEAMR RATVEREMEL RHKNEMLRVE AEARARAKAE RENADIIREQ IRLKAAEHRQ TVLESIRTAG TLFGEGFRAF; Positive Samples: HeLa, Jurkat, MCF7, HepG2, Mouse liver, Mouse kidney, Mouse brain; Cellular Location: Mitochondrion inner membrane, Mitochondrion matrix, Single-pass membrane protein, mitochondrion nucleoid.

ATAD3A is a member of the AAA ATPase family, a family of proteins that catalyze ATP into ADP and are involved in several cellular functions such as cell-cycle regulation, protein proteolysis and transport. The AAA ATPase family is characterized by a highly conserved AAA motif containing Walker homology sequences and imparting ATPase activity. Mitochondrial membrane proteins ATAD3A/B contribute to the stabilization of nucleoids and may participate in the transformation pathway and the chemosensitivity of oligodendrogliomas. The gene encoding ATAD3A/B/C maps to human chromosome 1 and has been suggested to be an anti-apoptotic marker.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATAD3A
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.31 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ATAD3A
Gene Accession No. Q925I1, Q9NVI7
Gene Alias 2400004H09Rik; AAA-ATPase TOB3; Atad3; ATAD3A; ATPase family AAA domain-containing protein 3; ATPase family AAA domain-containing protein 3A; ATPase family, AAA domain containing 3A; Kiaa1273; mKIAA1273; RP5-832C2.1; Tob3
Gene Symbols ATAD3A
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-240 of human ATAD3A (NP_0011640061).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 108888, 55210
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.