Learn More
Invitrogen™ ATAD3A Polyclonal Antibody

Description
A suggested positive control is Daudi cell lysate. PA5-21159 can be used with blocking peptide PEP-1273.
Specifications
Specifications
| Antigen | ATAD3A |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | ATAD3A |
| Gene Accession No. | Q3KRE0, Q925I1, Q9NVI7 |
| Gene Alias | 2400004H09Rik; AAA-ATPase TOB3; Atad3; ATAD3A; ATPase family AAA domain-containing protein 3; ATPase family AAA domain-containing protein 3A; ATPase family, AAA domain containing 3A; Kiaa1273; mKIAA1273; RP5-832C2.1; Tob3 |
| Gene Symbols | ATAD3A |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.