Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATAD3B Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP176587PCP
Description
ATAD3B Polyclonal antibody specifically detects ATAD3B in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
ATAD3B | |
Polyclonal | |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
AAA-ATPase TOB3, ATPase family AAA domain-containing protein 3B, ATPase family, AAA domain containing 3B, KIAA1273TOB3AAA-TOB3 | |
Antibody was raised against a 14 amino acid synthetic peptide near the carboxy terminus of human ATAD3B. The immunogen is located within amino acids 570 - 620 of ATAD3B. Amino Acid Squence: ETLTSWSLATDPSY | |
0.1 mL | |
Protein Phosphatase | |
83858 | |
Store at 4C in the dark. | |
IgG |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
PerCP | |
PBS | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction