Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Atad5 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10057495
Description
Atad5 Polyclonal specifically detects Atad5 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Atad5 | |
Polyclonal | |
Unconjugated | |
TBS and 0.1% BSA with 0.09% Sodium Azide | |
ATPase family AAA domain-containing protein 5, ATPase family, AAA domain containing 5, C17orf41, chromosome fragility associated gene 1, Chromosome fragility-associated gene 1 protein, ELG1, enhanced level of genomic instability 1 homolog, FLJ12735, FRAG1chromosome 17 open reading frame 41 | |
Rabbit | |
Affinity Purified | |
RUO | |
79915 | |
Store at 4C. Do not freeze. |
Western Blot, Immunoprecipitation | |
0.2 mg/mL | |
Western Blot 1:2000-1:10000, Immunoprecipitation 2 - 5 ug/mg lysate | |
NP_079133.3 | |
ATAD5 | |
The immunogen recognized by this antibody maps to a region between residue 775 and 825 of human ATPase family, AAA domain containing 5 using the numbering given in entry NP_079133.3 (GeneID 79915). | |
100 μL | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction