Learn More
Tocris Bioscience™ aTAG 4531

Description
Mouse DMPK properties are provided in the supplementary file (see below).
MTH1 can be expressed as a fusion with a target protein of interest using genome engineering techniques via CRISPR-mediated locus-specific knock-in. See protocol for more information.
Custom knock-in cell lines for the dTAG and aTAG platforms are available from our sister brand R&D Systems . Email TPD@bio-techne.com to enquire.
Specifications
Specifications
| Chemical Name or Material | N-Cyclopropyl-6-(4-((4-(4-(((2-(2,6-dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)oxy)methyl)-1 H-1,2,3-triazol-1-yl)butyl)carbamoyl)-3-fluorophenyl)-7-fluoro-4-(phenylamino)quinoline-3-carboxamide |
| CAS | 2412985-00-1 |
| Quantity | 5 mg |
| Target | TAG Degradation Platforms |
| Molecular Formula | C46H39F2N9O7 |
| Purity | 98% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.