Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATAT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569188
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569188 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569188 Supplier Invitrogen™ Supplier No. PA569188
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 93%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 93%; Rat: 100%.

This gene encodes a protein that localizes to clathrin-coated pits, where it acetylates alpha tubulin on lysine 40. This process may be important in microtubule growth, for instance during chemotaxis and the formation of cilium. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATAT1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ATAT1
Gene Accession No. Q6MG11
Gene Alias 0610011P08Rik; 2610008K08Rik; 2610110G12Rik; 3110080J08Rik; Acetyltransferase mec-17 homolog; acetyltransferase mec-17 homolog {ECO:0000255; alpha tubulin acetyltransferase 1; alpha-TAT; alpha-TAT {ECO:0000255; alpha-TAT1; alpha-TAT1 {ECO:0000255; alpha-tubulin N-acetyltransferase 1; alpha-tubulin N-acetyltransferase 1 {ECO:0000255; alpha-tubulin N-acetyltransferase; alpha-tubulin N-acetyltransferase 1; Atat1; C23H6orf134; C6orf134; HAMAP-Rule:MF_03130}; hypothetical protein FLJ13158; MEC17; Nbla00487; Novel DUF738 containing protein (2610110G12Rik); RGD1303066; TAT; TAT {ECO:0000255
Gene Symbols ATAT1
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of rat ATAT1.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 361789
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.