Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ATAT1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5112992
Description
ATAT1 Polyclonal Antibody for Western Blot, ELISA
This gene encodes a protein that localizes to clathrin-coated pits, where it acetylates alpha tubulin on lysine 40. This process may be important in microtubule growth, for instance during chemotaxis and the formation of cilium. Alternative splicing results in multiple transcript variants.
Specifications
| ATAT1 | |
| Polyclonal | |
| Unconjugated | |
| ATAT1 | |
| 0610011P08Rik; 2610008K08Rik; 2610110G12Rik; 3110080J08Rik; Acetyltransferase mec-17 homolog; acetyltransferase mec-17 homolog {ECO:0000255; alpha tubulin acetyltransferase 1; alpha-TAT; alpha-TAT {ECO:0000255; alpha-TAT1; alpha-TAT1 {ECO:0000255; alpha-tubulin N-acetyltransferase 1; alpha-tubulin N-acetyltransferase 1 {ECO:0000255; alpha-tubulin N-acetyltransferase; alpha-tubulin N-acetyltransferase 1; Atat1; C23H6orf134; C6orf134; HAMAP-Rule:MF_03130}; hypothetical protein FLJ13158; MEC17; Nbla00487; Novel DUF738 containing protein (2610110G12Rik); RGD1303066; TAT; TAT {ECO:0000255 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 79969 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 3.03 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q5SQI0 | |
| ATAT1 | |
| Recombinant Human Alpha-tubulin N-acetyltransferase 1 protein (194-238AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction