Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATAT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561340
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561340 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561340 Supplier Invitrogen™ Supplier No. PA561340
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EVEPLCILDF YIHESVQRHG HGRELFQYML QKERVEPHQL AIDRPSQKLL KFLNKHYNLE TTVPQVNNFV IFE Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 99%.

This gene encodes a protein that localizes to clathrin-coated pits, where it acetylates alpha tubulin on lysine 40. This process may be important in microtubule growth, for instance during chemotaxis and the formation of cilium. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATAT1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ATAT1
Gene Accession No. Q5SQI0
Gene Alias 0610011P08Rik; 2610008K08Rik; 2610110G12Rik; 3110080J08Rik; Acetyltransferase mec-17 homolog; acetyltransferase mec-17 homolog {ECO:0000255; alpha tubulin acetyltransferase 1; alpha-TAT; alpha-TAT {ECO:0000255; alpha-TAT1; alpha-TAT1 {ECO:0000255; alpha-tubulin N-acetyltransferase 1; alpha-tubulin N-acetyltransferase 1 {ECO:0000255; alpha-tubulin N-acetyltransferase; alpha-tubulin N-acetyltransferase 1; Atat1; C23H6orf134; C6orf134; HAMAP-Rule:MF_03130}; hypothetical protein FLJ13158; MEC17; Nbla00487; Novel DUF738 containing protein (2610110G12Rik); RGD1303066; TAT; TAT {ECO:0000255
Gene Symbols ATAT1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ATAT1. Recombinant protein control fragment (Product #RP-99605).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79969
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.