Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ataxin 1 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP145237
Description
Ataxin 1 Polyclonal specifically detects Ataxin 1 in Human, Porcine, Canine samples. It is validated for Peptide ELISA.Specifications
Ataxin 1 | |
Polyclonal | |
Peptide ELISA Detection limit 1:64000 | |
NP_000323.2 | |
ATXN1 | |
Peptide with sequence C-DLEVQQATHREASP corresponding to internal region according to NP_000323.2. | |
0.1 mg | |
Primary | |
Reported variant NP_000323.2 and NP_001121636.1 represent identical protein. | |
Store at -20C. Avoid freeze-thaw cycles. |
ELISA | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
ataxin 1, ataxin 1), ataxin-1, ATX1spinocerebellar ataxia 1 (olivopontocerebellar ataxia 1, autosomal dominant, SCA1D6S504E, Spinocerebellar ataxia type 1 protein | |
Goat | |
Affinity Purified | |
RUO | |
6310 | |
Human, Pig, Canine | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction