Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ataxin-10 Antibody, DyLight 680, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10057491FR
Description
Ataxin-10 Polyclonal antibody specifically detects Ataxin-10 in Human samples. It is validated for Western BlotSpecifications
Ataxin-10 | |
Polyclonal | |
Western Blot | |
ataxin 10, ataxin-10, Brain protein E46 homolog, E46L, FLJ37990, HUMEEP, SCA10spinocerebellar ataxia 10, Spinocerebellar ataxia type 10 protein | |
The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human Ataxin 10 (spinocerebellar ataxia 10) using the numbering given in entry NP_037368.1 (GeneID 25814). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
DyLight 680 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
25814 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction