Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ataxin-2 Antibody, DyLight 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10058797DL594
Description
Ataxin-2 Polyclonal antibody specifically detects Ataxin-2 in Human, Mouse samples. It is validated for Western BlotSpecifications
Ataxin-2 | |
Polyclonal | |
Western Blot | |
ataxin 2, ataxin 2), ataxin-2, ATX2spinocerebellar ataxia 2 (olivopontocerebellar ataxia 2, autosomal dominant, SCA2FLJ46772, Spinocerebellar ataxia type 2 protein, TNRC13, trinucleotide repeat containing 13, Trinucleotide repeat-containing gene 13 protein | |
The immunogen recognized by this antibody maps to a region between residue 1262 and 1312 of human ataxin 2 (spinocerebellar ataxia 2) using the numbering given in entry NP_002964.2 (GeneID 6311). | |
0.1 mL | |
Cellular Markers, Hematopoietic Stem Cell Markers, Stem Cell Markers | |
6311 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
DyLight 594 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction