Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ataxin-3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP287047
Description
Ataxin-3 Polyclonal specifically detects Ataxin-3 in Human samples. It is validated for Western Blot.Specifications
Ataxin-3 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
4287 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
ataxin 3, ATX3AT3, EC 3.4.19.12, EC 3.4.22, Machado-Joseph disease protein 1, MJD1, MJDolivopontocerebellar ataxia3, autosomal dominant, ataxin 3), olivopontocerebellar ataxia 3, Spinocerebellar ataxia type 3 protein | |
The immunogen is a synthetic peptide directed towards the N-terminal region of human Ataxin-3 | |
100 μL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction