Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ataxin 3 Recombinant Rabbit Monoclonal Antibody (HL1909)
GREENER_CHOICE

Catalog No. PIMA547219
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547219 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA547219 Supplier Invitrogen™ Supplier No. MA547219
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Store as concentrated solution. Centrifuge briefly prior to opening vial.

Machado-Joseph disease is an autosomal dominant neurologic disorder, and is now known to be the same as previously described spinocerebellar ataxia-3. MJD protein (Ataxin-3) contains (CAG)n repeats in the coding region, and the expansion of these repeats from the normal 13-36 to 68-79 is the cause of Machado-Joseph disease. There is a negative correlation between the age of onset and CAG repeat numbers. This protein interacts with key regulators (CBP, p300 and PCAF) of transcription and represses transcription, and also acts as a histone-binding protein that regulates transcription. MJD is a deubiquitinating enzyme.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ataxin 3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone HL1909
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene ATXN3
Gene Accession No. O35815, P54252, Q9CVD2
Gene Alias 2210008M02Rik; AI463012; AI647473; AT3; ataxin 3; ataxin 3 variant an; ataxin 3 variant ao; ataxin 3 variant at; ataxin 3 variant e; ataxin 3 variant h; ataxin 3 variant m; ataxin 3 variant r; ataxin 3 variant ref; ataxin 3 variant y; ataxin3; ataxin-3; ATX3; ATXN3; JOS; josephin; Machado-Joseph disease (spinocerebellar ataxia 3, olivopontocerebellar ataxia 3, autosomal dominant, ataxin 3); Machado-Joseph disease (spinocerebellar ataxia 3, olivopontocerebellar ataxia 3, autosomal dominant, ataxin 3) homolog; Machado-Joseph disease protein 1; Machado-Joseph disease protein 1 homolog; MJD; MJD1; olivopontocerebellar ataxia 3; Rsca3; Sca3; spin; Spinocerebellar ataxia type 3 protein
Gene Symbols ATXN3
Host Species Rabbit
Immunogen Recombinant fragment of human Ataxin 3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 110616, 4287, 60331
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.