Learn More
Invitrogen™ Ataxin 3 Polyclonal Antibody

Description
Ataxin 3 Polyclonal Antibody for Western Blot, IHC (P), Flow
Specifications
Specifications
| Antigen | Ataxin 3 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.45 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | ATXN3 |
| Gene Accession No. | P54252 |
| Gene Alias | 2210008M02Rik; AI463012; AI647473; AT3; ataxin 3; ataxin 3 variant an; ataxin 3 variant ao; ataxin 3 variant at; ataxin 3 variant e; ataxin 3 variant h; ataxin 3 variant m; ataxin 3 variant r; ataxin 3 variant ref; ataxin 3 variant y; ataxin3; ataxin-3; ATX3; ATXN3; JOS; josephin; Machado-Joseph disease (spinocerebellar ataxia 3, olivopontocerebellar ataxia 3, autosomal dominant, ataxin 3); Machado-Joseph disease (spinocerebellar ataxia 3, olivopontocerebellar ataxia 3, autosomal dominant, ataxin 3) homolog; Machado-Joseph disease protein 1; Machado-Joseph disease protein 1 homolog; MJD; MJD1; olivopontocerebellar ataxia 3; Rsca3; Sca3; spin; Spinocerebellar ataxia type 3 protein |
| Gene Symbols | ATXN3 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.