Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ataxin 7 Antibody, Alexa Fluor™ 750, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP142657AF750
Description
Ataxin 7 Polyclonal antibody specifically detects Ataxin 7 in Human samples. It is validated for Western BlotSpecifications
Ataxin 7 | |
Polyclonal | |
Western Blot | |
ADCAII, ataxin 7, FLJ17787, SCA7 | |
The immunogen for this product maps to a region between residue 842 and 892 of human spinocerebellar ataxia type 7 using the numbering given in entry NP_000324.1 (GeneID 6314). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
6314 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction