Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ataxin 7-Like 3 Antibody, DyLight 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299131V
Description
Ataxin 7-Like 3 Polyclonal specifically detects Ataxin 7-Like 3 in Human samples. It is validated for Western Blot.Specifications
Ataxin 7-Like 3 | |
Polyclonal | |
Western Blot | |
Ataxin-7-Like Protein 3, ATXN7L3, SAGA-Associated Factor 11 Homolog | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human Ataxin 7-Like 3. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
DyLight 405 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
56970 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction