Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ataxin-7 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501563090
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-3090 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-3090 Supplier Bethyl Laboratories, Inc Supplier No. A302638A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

Spinocerebellar ataxia type 7 (SCA7) belongs to a class of hereditary neurodegenrative diseases characterized by abnormally expanded polyglutamine (CAG) repeats at the N-terminus. This autosomal domit cerebellar ataxia primarily affects the cerebellum, retina, and brain stem and causes dementia, macular degeneration and other neurodegenerative characteristics. SCA7 gene encodes the ubiquitous 892 residue protein ataxin-7. This protein is typically located in the cytoplasm and on the nuclear membrane of normal brain neurons. In cells where there is a mutation of the SCA7 gene, ataxin 7 accumulates in intranuclear inclusions and can result in cell death.

Specifications

Antigen Ataxin-7
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene ATXN7
Gene Accession No. O15265
Gene Alias ADCAII, OPCA3, SCA7
Gene Symbols ATXN7
Host Species Rabbit
Immunogen Between 842 and 892
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6314
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.