Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATBF1 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS11805R
Description
Transcriptional repressor. It inhibits the enhancer element of the AFP gene by binding to its AT-rich core sequence. Regulator of myoblasts differentiation through the binding to the AT-rich sequence of MYF6 promoter and promoter repression. Down-regulates the MUC5AC promoter in gastric cancer. Miura Y., J. Biol. Chem. 270:26840-26848(1995). Morinaga T., Mol. Cell. Biol. 11:6041-6049(1991). Martin J., Nature 432:988-994(2004).Specifications
| ATBF1 | |
| Polyclonal | |
| Unconjugated | |
| ZFHX3 | |
| A230102L03Rik; alpha-fetoprotein enhancer binding protein; Alpha-fetoprotein enhancer-binding protein; AT motif binding factor 1; AT motif-binding factor 1; ATBF1; AT-binding transcription factor 1; ATBT; C16orf47; mKIAA4228; Putative uncharacterized protein C16orf47; WBP9; ZFH-3; ZFHX3; zinc finger homeobox 3; Zinc finger homeobox protein 3; Zinc finger homeodomain protein 3; ZNF927 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 463 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1 μg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
| Q15911 | |
| ZFHX3 | |
| KLH conjugated synthetic peptide derived from human ATBF1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction