Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ATCAY Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109423
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109423 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109423 Supplier Invitrogen™ Supplier No. PA5109423
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MGTTEATLRM ENVDVKEEWQ DEDLPRPLPE ETGVELLGSP VEDTSSPPNT LNFNGAHRKR KTLVAPEINI SLDQSEGSLL SDDFLDTPDD LDINVDDIET PDETD.

This gene encodes a neuron-restricted protein that contains a CRAL-TRIO motif common to proteins that bind small lipophilic molecules. Mutations in this gene are associated with cerebellar ataxia, Cayman type.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATCAY
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.78 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene ATCAY
Gene Accession No. Q1M168, Q8BHE3
Gene Alias 3322401A10Rik; ataxia cayman type protein; ataxia cerebellar Cayman type; ataxia, cerebellar, Cayman type; ataxia, cerebellar, Cayman type homolog (human); Atcay; ATCAY, caytaxin; BB077577; BNIP-2-homolgy; BNIP-2-homology; BNIP-H; Cayman ataxia; caytaxin; CLAC; hes; hesitant; ji; jittery; KIAA1872
Gene Symbols ATCAY
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-105 of human ATCAY (NP_149053.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16467, 362826
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.